missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Kestose, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 470-69-9 |
| Physical Form | Neat |
| Linear Formula | C18H32O16 |
| MDL Number | MFCD00142647 |
| Synonym | beta-D-Fruf-(2->1)-beta-D-Fruf-(2->1)-alpha-D-Glup |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 504.44 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Analytical Standard |