missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Isopropylpyrazole 98.0+%, TCI America™
$106.25 - $307.57
Chemical Identifiers
| CAS | 18952-87-9 |
|---|---|
| Molecular Formula | C6H10N2 |
| Molecular Weight (g/mol) | 110.16 |
| MDL Number | MFCD07468920 |
| InChI Key | ANMVTDVBEDVFRB-UHFFFAOYSA-N |
| PubChem CID | 13469816 |
| IUPAC Name | 1-(propan-2-yl)-1H-pyrazole |
| SMILES | CC(C)N1C=CC=N1 |
Chemical Identifiers
| 18952-87-9 | |
| 110.16 | |
| ANMVTDVBEDVFRB-UHFFFAOYSA-N | |
| 1-(propan-2-yl)-1H-pyrazole |
| C6H10N2 | |
| MFCD07468920 | |
| 13469816 | |
| CC(C)N1C=CC=N1 |
Specifications
| 18952-87-9 | |
| 142°C | |
| MFCD07468920 | |
| 1993 | |
| CC(C)N1C=CC=N1 | |
| 110.16 | |
| 110.16 | |
| Liquid |
| Colorless | |
| C6H10N2 | |
| 1 mL | |
| ANMVTDVBEDVFRB-UHFFFAOYSA-N | |
| 1-(propan-2-yl)-1H-pyrazole | |
| 13469816 | |
| ≥98.0% (GC,T) | |
| 1-Isopropylpyrazole |
Safety and Handling
TSCA : No