accessibility menu, dialog, popup

UserName

H3CCH3NN

CAS RN 18952-87-9

IUPAC Name: 1-(propan-2-yl)-1H-pyrazole
Synonyms: 1-isopropylpyrazole
Molecular Weight (g/mol): 110.16
Molecular Formula: C6H10N2
InChi Key: ANMVTDVBEDVFRB-UHFFFAOYSA-N
SMILES: CC(C)N1C=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results