missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Isopropylpyrazole 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 142°C |
| Quantity | 5 mL |
| UN Number | 1993 |
| Formula Weight | 110.16 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Isopropylpyrazole |
Safety and Handling
| TSCA | No |