missing translation for 'onlineSavingsMsg'
Learn More

1-Isopropenyl-2-benzimidazolidinone, 98+%

Catalog No. AAL1762606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1762606 5 g
AAL1762603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1762606 Supplier Thermo Scientific Chemicals Supplier No. L1762606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52099-72-6 | C10H10N2O | 174.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52099-72-6
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD00218253
InChI Key XFASJWLBXHWUMW-UHFFFAOYSA-N
Synonym 1-prop-1-en-2-yl-1h-benzo d imidazol-2 3h-one, 1-isopropenyl-2-benzimidazolidinone, 1,3-dihydro-1-1-methylethenyl-2h-benzimidazole-2-one, 1-isopropenylbenzimidazolone, 1,3-dihydro-1-1-methylvinyl-2h-benzimidazol-2-one, 1-prop-1-en-2-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-isopropenyl-1,3-dihydro-benzoimidazol-2-one, 1-isopropenyl-2-benzimidazolinone, 1-isopropenyl-1,3-dihydro-2h-1,3-benzimidazol-2-one, 1-isopropenyl-2,3-dihydro-1h-benzo d imidazol-2-one
PubChem CID 100278
IUPAC Name 3-prop-1-en-2-yl-1H-benzimidazol-2-one
SMILES CC(=C)N1C2=CC=CC=C2NC1=O

Specifications

Melting Point 121°C to 123°C
Quantity 5 g
Beilstein 140587
Formula Weight 174.2
Percent Purity ≥98%
Chemical Name or Material 1-Isopropenyl-2-benzimidazolidinone
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 257-661-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only