accessibility menu, dialog, popup

UserName

H3CCH2NOHN

CAS RN 52099-72-6

IUPAC Name: 1-(prop-1-en-2-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 1-(prop-1-en-2-yl)-3H-1,3-benzodiazol-2-one
Molecular Weight (g/mol): 174.2
Molecular Formula: C10H10N2O
InChi Key: XFASJWLBXHWUMW-UHFFFAOYSA-N
SMILES: CC(=C)N1C(=O)NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results