missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Isoindolinone, Thermo Scientific™
Chemical Identifiers
| CAS | 480-91-1 |
|---|---|
| Molecular Formula | C8H7NO |
| Molecular Weight (g/mol) | 133.15 |
| InChI Key | PXZQEOJJUGGUIB-UHFFFAOYSA-N |
| IUPAC Name | 2,3-dihydro-1H-isoindol-1-one |
| SMILES | O=C1NCC2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC465180010
|
Thermo Scientific Chemicals
465180010 |
1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC465180050
|
Thermo Scientific Chemicals
465180050 |
5 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Safety and Handling
GHS H Statement Causes serious eye irritation.
GHS P Statement Wear eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
RUO â Research Use Only