missing translation for 'onlineSavingsMsg'
Learn More

1-Isoindolinone

Catalog No. AC465180010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465180010 1 g
AC465180050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465180010 Supplier Thermo Scientific Chemicals Supplier No. 465180010

CAS: 480-91-1 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 480-91-1
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
InChI Key PXZQEOJJUGGUIB-UHFFFAOYSA-N
IUPAC Name 2,3-dihydro-1H-isoindol-1-one
SMILES O=C1NCC2=CC=CC=C12

Specifications

Melting Point 150.0°C to 160.0°C
Quantity 1 g
Formula Weight 133.15
Chemical Name or Material 1-Isoindolinone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes serious eye irritation.

Precautionary Statement GHS P Statement Wear eye protection/face protection.

Wash face, hands and any exposed skin thoroughly after handling.

If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only