missing translation for 'onlineSavingsMsg'
Learn More

1-(Isocyanomethyl)-1H-benzotriazole 97.0+%, TCI America™

Catalog No. I04551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04551G 1 g
I04555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04551G Supplier TCI America Supplier No. I04551G

Chemical Identifiers

CAS 87022-42-2
Molecular Formula C8H6N4
Molecular Weight (g/mol) 158.164
MDL Number MFCD00961633
InChI Key YZTNZXMSOPEFKC-UHFFFAOYSA-N
Synonym (1H-Benzotriazol-1-yl)methyl Isocyanide, 1-(Isocyanidomethyl)-1H-benzotriazole
PubChem CID 3832111
IUPAC Name 1-(isocyanomethyl)benzotriazole
SMILES [C-]#[N+]CN1C2=CC=CC=C2N=N1

Specifications

Quantity 1 g
Formula Weight 158.16
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Isocyanomethyl)-1H-benzotriazole