accessibility menu, dialog, popup

UserName

CAS RN 87022-42-2

IUPAC Name: 1-(isocyanomethyl)-1H-1,2,3-benzotriazole
Synonyms: 1-(isocyanomethyl)-1,2,3-benzotriazole
Molecular Weight (g/mol): 158.16
Molecular Formula: C8H6N4
InChi Key: YZTNZXMSOPEFKC-UHFFFAOYSA-N
SMILES: [C-]#[N+]CN1N=NC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results