missing translation for 'onlineSavingsMsg'
Learn More

1-Isocyanoadamantane 97.0+%, TCI America™

Catalog No. I08241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08241G 1 g
I08245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08241G Supplier TCI America Supplier No. I08241G

Chemical Identifiers

CAS 22110-53-8
Molecular Formula C11H15N
Molecular Weight (g/mol) 161.25
MDL Number MFCD02258889
InChI Key RPRJRJNIFAYHRF-UHFFFAOYSA-N
Synonym 1-Adamantyl Isocyanide
PubChem CID 140885
IUPAC Name 1-isocyanoadamantane
SMILES [C-]#[N+]C12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 197°C
Color Yellow
Quantity 1 g
Formula Weight 161.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Isocyanoadamantane