missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Iodopentane (stabilized with Copper chip) 98.0+%, TCI America™
$98.18 - $721.93
Chemical Identifiers
| CAS | 628-17-1 |
|---|---|
| Molecular Formula | C5H11I |
| Molecular Weight (g/mol) | 198.047 |
| MDL Number | MFCD00001100 |
| InChI Key | BLXSFCHWMBESKV-UHFFFAOYSA-N |
| Synonym | n-amyl iodide, amyl iodide, pentyl iodide, n-pentyl iodide, pentane, 1-iodo, 1-pentyl iodide, 1-jodpentan, 1-jodpentan czech, iodopentane, pentyliodide |
| PubChem CID | 12335 |
| IUPAC Name | 1-iodopentane |
| SMILES | CCCCCI |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I006625ML
|
TCI America
I006625ML |
25 mL |
Each for $98.18
|
|
|||||
|
I0066250ML
|
TCI America
I0066250ML |
250 mL |
Each for $721.93
|
|
|||||
Chemical Identifiers
| 628-17-1 | |
| 198.047 | |
| BLXSFCHWMBESKV-UHFFFAOYSA-N | |
| 12335 | |
| CCCCCI |
| C5H11I | |
| MFCD00001100 | |
| n-amyl iodide, amyl iodide, pentyl iodide, n-pentyl iodide, pentane, 1-iodo, 1-pentyl iodide, 1-jodpentan, 1-jodpentan czech, iodopentane, pentyliodide | |
| 1-iodopentane |
Specifications
| 628-17-1 | |
| 155°C | |
| MFCD00001100 | |
| 1993 | |
| BLXSFCHWMBESKV-UHFFFAOYSA-N | |
| 1-iodopentane | |
| 12335 | |
| ≥98.0% (GC) | |
| 1-Iodopentane (stabilized with Copper chip) |
| Yellow | |
| C5H11I | |
| 25 mL | |
| n-amyl iodide, amyl iodide, pentyl iodide, n-pentyl iodide, pentane, 1-iodo, 1-pentyl iodide, 1-jodpentan, 1-jodpentan czech, iodopentane, pentyliodide | |
| CCCCCI | |
| 198.047 | |
| 198.05 | |
| Liquid |
Safety and Handling
RTECSNumber : SA2975600
TSCA : Yes