missing translation for 'onlineSavingsMsg'
Learn More

1-Iodopentane, 98%, stab with copper

CAS: 628-17-1 | C5H11I | 198.047 g/mol

Supplier:  Thermo Scientific Chemicals B2078136

 View more versions of this product

Catalog No. AAB2078136

Only null left
Explore available promotions

Description

Description

1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in dark place. Incompatible with strong oxidizing agents.
Specifications

Specifications

1-Iodopentane
628-17-1
1.515
51°C (123°F)
C5H11I
CH3(CH2)4I
500 g
1730982
n-amyl iodide, amyl iodide, pentyl iodide, n-pentyl iodide, pentane, 1-iodo, 1-pentyl iodide, 1-jodpentan, 1-jodpentan czech, iodopentane, pentyliodide
BLXSFCHWMBESKV-UHFFFAOYSA-N
1-iodopentane
12335
98%
stabilized with copper
-86°C
154°C to 156°C
Characteristic
1.495
MFCD00001100
UN1993
Light sensitive
Insoluble in water.
CCCCCI
198.047
198.05

Chemical Identifiers

628-17-1
198.047
BLXSFCHWMBESKV-UHFFFAOYSA-N
12335
CCCCCI
C5H11I
MFCD00001100
n-amyl iodide, amyl iodide, pentyl iodide, n-pentyl iodide, pentane, 1-iodo, 1-pentyl iodide, 1-jodpentan, 1-jodpentan czech, iodopentane, pentyliodide
1-iodopentane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 211-030-0

RTECSNumber : SA2975600

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only