Learn More
1-Iodopentane, 98%, stab with copper
CAS: 628-17-1 | C5H11I | 198.047 g/mol
Supplier: Thermo Scientific Chemicals B2078122
Description
1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Iodopentane (pentyl iodide) was used in the preparation of 1-pentyl radical by shock heating process.
Solubility
Insoluble in water.
Notes
Light sensitive. Store in dark place. Incompatible with strong oxidizing agents.
Specifications
| 1-Iodopentane | |
| 628-17-1 | |
| 1.515 | |
| 51°C (123°F) | |
| C5H11I | |
| CH3(CH2)4I | |
| 100 g | |
| 1730982 | |
| n-amyl iodide, amyl iodide, pentyl iodide, n-pentyl iodide, pentane, 1-iodo, 1-pentyl iodide, 1-jodpentan, 1-jodpentan czech, iodopentane, pentyliodide | |
| BLXSFCHWMBESKV-UHFFFAOYSA-N | |
| 1-iodopentane | |
| 12335 | |
| 98% |
| stabilized with copper | |
| -86°C | |
| 154°C to 156°C | |
| Characteristic | |
| 1.495 | |
| MFCD00001100 | |
| UN1993 | |
| Light sensitive | |
| Insoluble in water. | |
| CCCCCI | |
| 198.047 | |
| 198.05 |
Chemical Identifiers
| 628-17-1 | |
| 198.047 | |
| BLXSFCHWMBESKV-UHFFFAOYSA-N | |
| 12335 | |
| CCCCCI |
| C5H11I | |
| MFCD00001100 | |
| n-amyl iodide, amyl iodide, pentyl iodide, n-pentyl iodide, pentane, 1-iodo, 1-pentyl iodide, 1-jodpentan, 1-jodpentan czech, iodopentane, pentyliodide | |
| 1-iodopentane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 211-030-0
RTECSNumber : SA2975600
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only