Learn More
1-Iodooctane, 98+%, stab. with copper
CAS: 629-27-6 | C8H17I | 240.128 g/mol
Supplier: Thermo Scientific Chemicals B2221422
Description
Alkylation of [2,2?-([2,2?-bipyridine]-6,6?-diyl)bis[phenolato]-N,N?,O,O?]nickel(II)was performed using the catalytic reduction of 1-iodooctane. Primary alkyl iodide 1-iodooctane reacts with lithium aluminum deuteride (LAD) in a 1:0.2 ratio to form 1-deuteriooctane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAlkylation of [2,2′-([2,2′-bipyridine]-6,6′-diyl)bis[phenolato]-N,N′,O,O′]nickel(II)was performed using the catalytic reduction of 1-iodooctane. Primary alkyl iodide 1-iodooctane reacts with lithium aluminum deuteride (LAD) in a 1:0.2 ratio to form 1-deuteriooctane.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, bases.
Specifications
| 1-Iodooctane | |
| 629-27-6 | |
| 1.325 | |
| 95°C (203°F) | |
| 1.4878 | |
| MFCD00001105 | |
| 1697479 | |
| n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctane | |
| UWLHSHAHTBJTBA-UHFFFAOYSA-N | |
| 1-iodooctane | |
| 12380 | |
| ≥98% |
| stabilized with copper | |
| -46°C | |
| 225°C to 226°C | |
| C8H17I | |
| CH3(CH2)7I | |
| 100 g | |
| Light sensitive | |
| Insoluble in water. | |
| CCCCCCCCI | |
| 240.128 | |
| 240.13 |
Chemical Identifiers
| 629-27-6 | |
| 240.128 | |
| UWLHSHAHTBJTBA-UHFFFAOYSA-N | |
| 12380 | |
| CCCCCCCCI |
| C8H17I | |
| MFCD00001105 | |
| n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctane | |
| 1-iodooctane |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 211-084-5
RTECSNumber : RG9700100
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only