missing translation for 'onlineSavingsMsg'
Learn More

1-Iodooctane, 98+%, stab. with copper

Catalog No. p-7026286
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2221414 25 g
AAB2221422 100 g
AAB2221436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2221414 Supplier Thermo Scientific Chemicals Supplier No. B2221414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 629-27-6 | C8H17I | 240.128 g/mol

Alkylation of [2,2?-([2,2?-bipyridine]-6,6?-diyl)bis[phenolato]-N,N?,O,O?]nickel(II)was performed using the catalytic reduction of 1-iodooctane. Primary alkyl iodide 1-iodooctane reacts with lithium aluminum deuteride (LAD) in a 1:0.2 ratio to form 1-deuteriooctane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alkylation of [2,2′-([2,2′-bipyridine]-6,6′-diyl)bis[phenolato]-N,N′,O,O′]nickel(II)was performed using the catalytic reduction of 1-iodooctane. Primary alkyl iodide 1-iodooctane reacts with lithium aluminum deuteride (LAD) in a 1:0.2 ratio to form 1-deuteriooctane.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, bases.

Chemical Identifiers

CAS 629-27-6
Molecular Formula C8H17I
Molecular Weight (g/mol) 240.128
MDL Number MFCD00001105
InChI Key UWLHSHAHTBJTBA-UHFFFAOYSA-N
Synonym n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctane
PubChem CID 12380
IUPAC Name 1-iodooctane
SMILES CCCCCCCCI

Specifications

Melting Point -46°C
Density 1.325
Boiling Point 225°C to 226°C
Flash Point 95°C (203°F)
Refractive Index 1.4878
Linear Formula CH3(CH2)7I
Quantity 25 g
Beilstein 1697479
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 240.13
Percent Purity ≥98%
Chemical Name or Material 1-Iodooctane, stabilized with copper
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 211-084-5
RTECSNumber RG9700100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only