missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Iodohexane (stabilized with Copper chip) 97.0+%, TCI America™
$128.62 - $365.35
Chemical Identifiers
| CAS | 638-45-9 |
|---|---|
| Molecular Formula | C6H13I |
| Molecular Weight (g/mol) | 212.074 |
| MDL Number | MFCD00001102 |
| InChI Key | ANOOTOPTCJRUPK-UHFFFAOYSA-N |
| PubChem CID | 12527 |
| IUPAC Name | 1-iodohexane |
| SMILES | CCCCCCI |
Chemical Identifiers
| 638-45-9 | |
| 212.074 | |
| ANOOTOPTCJRUPK-UHFFFAOYSA-N | |
| 1-iodohexane |
| C6H13I | |
| MFCD00001102 | |
| 12527 | |
| CCCCCCI |
Specifications
| 638-45-9 | |
| 180°C | |
| MFCD00001102 | |
| 3082 | |
| ANOOTOPTCJRUPK-UHFFFAOYSA-N | |
| 1-iodohexane | |
| 12527 | |
| ≥97.0% (GC) | |
| 1-Iodohexane (stabilized with Copper chip) |
| Yellow | |
| C6H13I | |
| 25 g | |
| hexyl iodide, n-hexyl iodide, hexane, 1-iodo, 1-hexyl iodide, iodohexane, 1-iodo-hexane, hexane,1-iodo, n-hexyliodide, iodo-hexane, 1-iodanylhexane | |
| CCCCCCI | |
| 212.074 | |
| 212.07 | |
| Liquid |
Safety and Handling
TSCA : Yes