missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-4-(trifluoromethoxy)benzene 98.0+%, TCI America™

Catalog No. T21535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T21535G 5 g
T215325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T21535G Supplier TCI America Supplier No. T21535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103962-05-6
Molecular Formula C7H4F3IO
Molecular Weight (g/mol) 288.01
MDL Number MFCD00042411
InChI Key RTUDBROGOZBBIC-UHFFFAOYSA-N
Synonym 1-iodo-4-trifluoromethoxy benzene, 4-trifluoromethoxy iodobenzene, 4-iodotrifluoromethoxybenzene, 1-iado-4-trifluoromethoxy benzene, p-trifluoromethoxy iodobenzene, 4-trifluoromethoxy-iodobenzene, 1-iodo-4-trifluoromethoxy-benzene, benzene, 1-iodo-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4-iodoanisole, 4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene
PubChem CID 2777294
IUPAC Name 1-iodo-4-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=C(I)C=C1

Specifications

Boiling Point 97°C
Quantity 5 g
Formula Weight 288.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodo-4-(trifluoromethoxy)benzene