missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-4-(trifluoromethoxy)benzene, 98%

Catalog No. AAB2362803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2362803 1 g
AAB2362806 5 g
AAB2362814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2362803 Supplier Thermo Scientific Chemicals Supplier No. B2362803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103962-05-6 | C7H4F3IO | 288.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103962-05-6
Molecular Formula C7H4F3IO
Molecular Weight (g/mol) 288.01
MDL Number MFCD00042411
InChI Key RTUDBROGOZBBIC-UHFFFAOYSA-N
Synonym 1-iodo-4-trifluoromethoxy benzene, 4-trifluoromethoxy iodobenzene, 4-iodotrifluoromethoxybenzene, 1-iado-4-trifluoromethoxy benzene, p-trifluoromethoxy iodobenzene, 4-trifluoromethoxy-iodobenzene, 1-iodo-4-trifluoromethoxy-benzene, benzene, 1-iodo-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4-iodoanisole, 4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene
PubChem CID 2777294
IUPAC Name 1-iodo-4-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=C(I)C=C1

Specifications

Density 1.909
Boiling Point 72°C to 74°C (15 mmHg)
Flash Point 77°C (170°F)
Refractive Index 1.5033
Quantity 1 g
Beilstein 6193794
Sensitivity Light sensitive
Formula Weight 288.01
Percent Purity 98%
Chemical Name or Material 1-Iodo-4-(trifluoromethoxy)benzene
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only