missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-4-nitrobenzene 99.0+%, TCI America™

Catalog No. p-7131188
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
I006425G 25 g
I0064100G 100 g
2 options

Catalog No. I006425G

Supplier: TCI America I006425G

Only null left

Chemical Identifiers

CAS 636-98-6
Molecular Formula C6H4INO2
Molecular Weight (g/mol) 249.007
MDL Number MFCD00007299
InChI Key SCCCFNJTCDSLCY-UHFFFAOYSA-N
Synonym benzene, 1-iodo-4-nitro, p-iodonitrobenzene, 4-iodonitrobenzene, p-nitroiodobenzene, 4-nitroiodobenzene, 4-iodo-nitrobenzene, p-nitrophenyl iodide, 1-iodo-4-nitro-benzene, 4-iodo-1-nitrobenzene, 4-nitro-1-iodobenzene
PubChem CID 12495
IUPAC Name 1-iodo-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])I

Specifications

Melting Point 173°C
Quantity 25 g
Formula Weight 249.01
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Iodo-4-nitrobenzene