missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-4-n-pentylbenzene, 98%

Catalog No. AAH2735603
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2735603 1 g
AAH2735609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2735603 Supplier Thermo Scientific Chemicals Supplier No. H2735603

CAS: 85017-60-3 | C11H15I | 274.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85017-60-3
Molecular Formula C11H15I
Molecular Weight (g/mol) 274.145
MDL Number MFCD00079767
InChI Key MTRMOYSZRPLAOL-UHFFFAOYSA-N
Synonym 4-iodopentylbenzene, 1-iodo-4-n-pentylbenzene, 4-pentyliodobenzene, #, 1-iodanyl-4-pentyl-benzene
PubChem CID 145057
IUPAC Name 1-iodo-4-pentylbenzene
SMILES CCCCCC1=CC=C(C=C1)I

Specifications

Density 1.38
Boiling Point 88°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 274.14
Percent Purity 98%
Chemical Name or Material 1-Iodo-4-n-pentylbenzene
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only