missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-4-isopropylbenzene, 97%

Catalog No. AAB2547614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2547614 25 g
AAB2547606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2547614 Supplier Thermo Scientific Chemicals Supplier No. B2547614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17356-09-1 | C9H11I | 246.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17356-09-1
Molecular Formula C9H11I
Molecular Weight (g/mol) 246.091
MDL Number MFCD00013711
InChI Key PQJOSEVTIKYWLH-UHFFFAOYSA-N
Synonym 1-iodo-4-isopropylbenzene, p-iodocumene, 4-iodoisopropylbenzene, 2-4'-iodophenyl propane, 4-iodocumene, p-iodoisopropylbenzene, 1-iodo-4-propan-2-yl benzene, 4-isopropyliodobenzene, benzene,1-iodo-4-1-methylethyl, benzene, 1-iodo-4-1-methylethyl
PubChem CID 87073
IUPAC Name 1-iodo-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(C=C1)I

Specifications

Density 1.525
Boiling Point 236°C to 238°C
Refractive Index 1.578
Quantity 25 g
Beilstein 2205506
Sensitivity Light sensitive
Formula Weight 246.09
Percent Purity 97%
Chemical Name or Material 1-Iodo-4-isopropylbenzene
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 241-384-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only