missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-3-(trifluoromethoxy)benzene 98.0+%, TCI America™

Catalog No. I08135G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I08135G Supplier TCI America Supplier No. I08135G

Chemical Identifiers

CAS 198206-33-6
Molecular Formula C7H4F3IO
Molecular Weight (g/mol) 288.01
MDL Number MFCD01090992
InChI Key UQZXQSQWKJZHCD-UHFFFAOYSA-N
Synonym 1-iodo-3-trifluoromethoxy benzene, 3-trifluoromethoxy iodobenzene, 3-iodo-1-trifluoromethoxy benzene, m-iodotrifluoromethoxybenzene, 1-iodo-3-trifluoromethoxy-benzene, benzene, 1-iodo-3-trifluoromethoxy, pubchem4438, acmc-1c5ss, 3-trifluoromethoxyiodobenzene, ksc494i7t
PubChem CID 2777293
IUPAC Name 1-iodo-3-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=CC(I)=C1

Specifications

Boiling Point 71°C
Quantity 5 g
Formula Weight 288.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodo-3-(trifluoromethoxy)benzene