missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-3-nitrobenzene, 99%

Catalog No. AAB2160514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2160514 25 g
AAB2160522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2160514 Supplier Thermo Scientific Chemicals Supplier No. B2160514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 645-00-1 | C6H4INO2 | 249.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 645-00-1
Molecular Formula C6H4INO2
Molecular Weight (g/mol) 249.007
MDL Number MFCD00007218
InChI Key CBYAZOKPJYBCHE-UHFFFAOYSA-N
Synonym 3-iodonitrobenzene, m-iodonitrobenzene, benzene, 1-iodo-3-nitro, 3-nitroiodobenzene, meta-iodonitrobenzene, 3-i-nitrobenzene, iodo-3-nitrobenzene, meta-nitroiodobenzene, 1-iodo-3-nitro-benzene, 3-nitro-iodobenzen
PubChem CID 12574
ChEBI CHEBI:67123
IUPAC Name 1-iodo-3-nitrobenzene
SMILES C1=CC(=CC(=C1)I)[N+](=O)[O-]

Specifications

Melting Point 35°C to 38°C
Boiling Point 279°C to 280°C
Flash Point 71°C (159°F)
Quantity 25 g
UN Number UN2926
Beilstein 1525167
Sensitivity Light sensitive
Formula Weight 249.01
Percent Purity 99%
Chemical Name or Material 1-Iodo-3-nitrobenzene
Hazard Category H228-H302+H312+H332-H319-H373
Hazard Statement GHS H Statement
H331-H228-H373-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable solid.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, TOXIC, ORGANIC, N.O.S.
EINECSNumber 211-427-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only