missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2-nitrobenzene 99.0+%, TCI America™

Catalog No. I006225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I006225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I006225G Supplier TCI America Supplier No. I006225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 609-73-4
Molecular Formula C6H4INO2
Molecular Weight (g/mol) 249.01
MDL Number MFCD00007088
InChI Key JXMZUNPWVXQADG-UHFFFAOYSA-N
Synonym 2-iodonitrobenzene, o-iodonitrobenzene, o-nitroiodobenzene, benzene, 1-iodo-2-nitro, 2-nitroiodobenzene, 2-iodo-1-nitrobenzene, 1-iodo-2-nitro-benzene, 2-iodo-nitrobenzene, 2-nitro-iodobenzene, benzene, iodonitro
PubChem CID 69115
IUPAC Name 1-iodo-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC=C1I

Specifications

Melting Point 52°C
Boiling Point 163°C
Quantity 25 g
Formula Weight 249.01
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Iodo-2-nitrobenzene