missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2-nitrobenzene, 97%

Catalog No. AAA1210014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1210014 25 g
AAA1210022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1210014 Supplier Thermo Scientific Chemicals Supplier No. A1210014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 609-73-4 | C6H4INO2 | 249.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 609-73-4
Molecular Formula C6H4INO2
Molecular Weight (g/mol) 249.01
MDL Number MFCD00007088
InChI Key JXMZUNPWVXQADG-UHFFFAOYSA-N
Synonym 2-iodonitrobenzene, o-iodonitrobenzene, o-nitroiodobenzene, benzene, 1-iodo-2-nitro, 2-nitroiodobenzene, 2-iodo-1-nitrobenzene, 1-iodo-2-nitro-benzene, 2-iodo-nitrobenzene, 2-nitro-iodobenzene, benzene, iodonitro
PubChem CID 69115
IUPAC Name 1-iodo-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC=C1I

Specifications

Melting Point 48°C to 52°C
Boiling Point 288°C to 289°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 2044110
Sensitivity Light sensitive
Formula Weight 249.01
Percent Purity 97%
Chemical Name or Material 1-Iodo-2-nitrobenzene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-200-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only