missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2-methylpropane (stabilized with Copper chip) 97.0+%, TCI America™

Catalog No. p-7131333
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I032225ML 25 mL
I0322250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I032225ML Supplier TCI America Supplier No. I032225ML
Only null left

Chemical Identifiers

CAS 513-38-2
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001084
InChI Key BTUGGGLMQBJCBN-UHFFFAOYSA-N
Synonym isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane
PubChem CID 10556
IUPAC Name 1-iodo-2-methylpropane
SMILES CC(C)CI

Specifications

Boiling Point 121°C
Quantity 25 mL
UN Number 2391
Formula Weight 184.02
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodo-2-methylpropane (stabilized with Copper chip)
RTECSNumber TZ4250000
TSCA Yes