accessibility menu, dialog, popup

UserName

CAS RN 513-38-2

IUPAC Name: 1-iodo-2-methylpropane
Synonyms: 1-iodo-2-methylpropane
Molecular Weight (g/mol): 184.02
Molecular Formula: C4H9I
InChi Key: BTUGGGLMQBJCBN-UHFFFAOYSA-N
SMILES: CC(C)CI
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)