Learn More
1-Iodo-2-methylpropane, 97%, stabilized
CAS: 513-38-2 | C4H9I | 184.02 g/mol
$149.62 - $149.62
Chemical Identifiers
| CAS | 513-38-2 |
|---|---|
| Molecular Formula | C4H9I |
| Molecular Weight (g/mol) | 184.02 |
| MDL Number | MFCD00001084 |
| InChI Key | BTUGGGLMQBJCBN-UHFFFAOYSA-N |
| Synonym | isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane |
| PubChem CID | 10556 |
| IUPAC Name | 1-iodo-2-methylpropane |
| SMILES | CC(C)CI |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC223740250
|
Thermo Scientific Chemicals
223740250 |
25 mL | Glass bottle |
Each for $149.62
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 513-38-2 | |
| 184.02 | |
| BTUGGGLMQBJCBN-UHFFFAOYSA-N | |
| 10556 | |
| CC(C)CI |
| C4H9I | |
| MFCD00001084 | |
| isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane | |
| 1-iodo-2-methylpropane |
Specifications
| 513-38-2 | |
| 100.0 | |
| Colorless to Orange | |
| 120.0°C to 121.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4948 to 1.4968 | |
| MFCD00001084 | |
| 1.59 | |
| isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane | |
| BTUGGGLMQBJCBN-UHFFFAOYSA-N | |
| 1-iodo-2-methylpropane | |
| copper | |
| 184.02 | |
| Liquid |
| 96.0 | |
| -93.0°C | |
| 1.5900g/mL | |
| 12°C | |
| 96% min. (GC) | |
| C4H9I | |
| (CH3)2CHCH2I | |
| 25 mL | |
| 15,5189 | |
| Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether | |
| CC(C)CI | |
| 184.02 | |
| 10556 | |
| 97% | |
| 1-Iodo-2-methylpropane, 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
EINECSNumber : 208-160-5
RTECSNumber : TZ4250000
TSCA : TSCA
RUO – Research Use Only