missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2-methylpropane, 97%, stab. with copper

Catalog No. p-7022904
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1588314 25 g
AAA1588322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1588314 Supplier Thermo Scientific Chemicals Supplier No. A1588314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 513-38-2 | C4H9I | 184.02 g/mol

1-Iodo-2-methylpropane is used as solvents, analytical reagents and organic synthesis of raw materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodo-2-methylpropane is used as solvents, analytical reagents and organic synthesis of raw materials.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light sensitive.

Chemical Identifiers

CAS 513-38-2
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001084
InChI Key BTUGGGLMQBJCBN-UHFFFAOYSA-N
Synonym isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane
PubChem CID 10556
IUPAC Name 1-iodo-2-methylpropane
SMILES CC(C)CI

Specifications

Melting Point -93°C
Density 1.599
Boiling Point 120°C to 121°C
Flash Point 12°C (53°F)
Refractive Index 1.496
Quantity 25 g
UN Number UN2391
Beilstein 1730927
Sensitivity Light sensitive
Merck Index 14,5144
Solubility Information Insoluble in water.
Formula Weight 184.02
Percent Purity 98%
Chemical Name or Material 1-Iodo-2-methylpropane, stabilized with copper
Show More Show Less
Hazard Category H225-H315-H319-H332-H335
Hazard Statement GHS H Statement
H225-H332-H315-H319-H335
Highly flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: IODOMETHYLPROPANES
EINECSNumber 208-160-5
RTECSNumber TZ4250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only