missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2,4-dinitrobenzene, 98%

Catalog No. p-7027135
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2365906 5 g
AAB2365914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2365906 Supplier Thermo Scientific Chemicals Supplier No. B2365906

CAS: 709-49-9 | C6H3IN2O4 | 294.004 g/mol

It is employed in the cloning and characterization of a biotic-stress-inducible glutathione transferase from Phaseolus vulgaris.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the cloning and characterization of a biotic-stress-inducible glutathione transferase from Phaseolus vulgaris.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from oxidizing agents, bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under dark.

Chemical Identifiers

CAS 709-49-9
Molecular Formula C6H3IN2O4
Molecular Weight (g/mol) 294.004
MDL Number MFCD00039738
InChI Key FXMKXMJLXRTQSW-UHFFFAOYSA-N
Synonym 2,4-dinitroiodobenzene, benzene, 1-iodo-2,4-dinitro, unii-not697x2xm, ccris 7907, 4-iodo-1,3-dintrobenzene, 6-iodo-1,3-dintrobenzene, 2,4-dinitro-1-iodobenzene, not697x2xm, 1-iodo-2,4-dinitro-benzene, 1-iod-2,4-dinitrobenzol
PubChem CID 69730
ChEBI CHEBI:59000
IUPAC Name 1-iodo-2,4-dinitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])I

Specifications

Melting Point 87°C to 90°C
Quantity 5 g
Beilstein 1113584
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 294.01
Percent Purity 98%
Chemical Name or Material 1-Iodo-2,4-dinitrobenzene
Hazard Category H315-H319-H334-H335
Hazard Statement GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P285-P302+P352-P304+P340-P305+P351+P338-P332+P313-P342+P311-P362-P501c
EINECSNumber 211-908-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only