missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2,4-dimethoxybenzene, 97%

CAS: 20469-63-0 | C8H9IO2 | 264.062 g/mol

$43.78 - $100.45

Chemical Identifiers

CAS 20469-63-0
Molecular Formula C8H9IO2
Molecular Weight (g/mol) 264.062
MDL Number MFCD00059268
InChI Key ZDUYJCKEORTAQE-UHFFFAOYSA-N
Synonym 2,4-dimethoxyiodobenzene, 2,4-dimethoxy-1-iodobenzene, 1,3-dimethoxy-4-iodobenzene, 1-iodo-2,4-dimethoxy-benzene, benzene, 1-iodo-2,4-dimethoxy, 4-iodo-1,3-dimethoxybenzene, pubchem3053, 4-iodo-3-methoxy anisole, dimethoxyiodobenzene, cambridge id 5107587
PubChem CID 140694
IUPAC Name 1-iodo-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)I)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1942003
View Documents
Thermo Scientific Chemicals
L1942003
1 g
Each for $43.78
Only null left
 
AAL1942006
View Documents
Thermo Scientific Chemicals
L1942006
5 g
Each for $100.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

20469-63-0
264.062
ZDUYJCKEORTAQE-UHFFFAOYSA-N
140694
COC1=CC(=C(C=C1)I)OC
C8H9IO2
MFCD00059268
2,4-dimethoxyiodobenzene, 2,4-dimethoxy-1-iodobenzene, 1,3-dimethoxy-4-iodobenzene, 1-iodo-2,4-dimethoxy-benzene, benzene, 1-iodo-2,4-dimethoxy, 4-iodo-1,3-dimethoxybenzene, pubchem3053, 4-iodo-3-methoxy anisole, dimethoxyiodobenzene, cambridge id 5107587
1-iodo-2,4-dimethoxybenzene

Specifications

20469-63-0
135°C (14 mmHg)
C8H9IO2
MFCD00059268
1945135
2,4-dimethoxyiodobenzene, 2,4-dimethoxy-1-iodobenzene, 1,3-dimethoxy-4-iodobenzene, 1-iodo-2,4-dimethoxy-benzene, benzene, 1-iodo-2,4-dimethoxy, 4-iodo-1,3-dimethoxybenzene, pubchem3053, 4-iodo-3-methoxy anisole, dimethoxyiodobenzene, cambridge id 5107587
COC1=CC(=C(C=C1)I)OC
264.062
264.06
1-Iodo-2,4-dimethoxybenzene
37°C to 41°C
>110°C (230°F)
1.61
1 g
Light sensitive
ZDUYJCKEORTAQE-UHFFFAOYSA-N
1-iodo-2,4-dimethoxybenzene
140694
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only