missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2,4-dimethoxybenzene, 97%

Catalog No. AAL1942006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1942006 5 g
AAL1942003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1942006 Supplier Thermo Scientific Chemicals Supplier No. L1942006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 20469-63-0 | C8H9IO2 | 264.062 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20469-63-0
Molecular Formula C8H9IO2
Molecular Weight (g/mol) 264.062
MDL Number MFCD00059268
InChI Key ZDUYJCKEORTAQE-UHFFFAOYSA-N
Synonym 2,4-dimethoxyiodobenzene, 2,4-dimethoxy-1-iodobenzene, 1,3-dimethoxy-4-iodobenzene, 1-iodo-2,4-dimethoxy-benzene, benzene, 1-iodo-2,4-dimethoxy, 4-iodo-1,3-dimethoxybenzene, pubchem3053, 4-iodo-3-methoxy anisole, dimethoxyiodobenzene, cambridge id 5107587
PubChem CID 140694
IUPAC Name 1-iodo-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)I)OC

Specifications

Melting Point 37°C to 41°C
Boiling Point 135°C (14 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.61
Quantity 5 g
Beilstein 1945135
Sensitivity Light sensitive
Formula Weight 264.06
Percent Purity 97%
Chemical Name or Material 1-Iodo-2,4-dimethoxybenzene
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only