missing translation for 'onlineSavingsMsg'
Learn More

1-Indanone 98.0+%, TCI America™

Catalog No. p-7131156
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
I00135G 5 g
I001325G 25 g
2 options

Catalog No. I00135G

Supplier: TCI America I00135G

Only null left

Chemical Identifiers

CAS 83-33-0
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00003785
InChI Key QNXSIUBBGPHDDE-UHFFFAOYSA-N
Synonym 1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone
PubChem CID 6735
ChEBI CHEBI:17404
IUPAC Name 2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=CC=CC=C21

Specifications

Melting Point 41°C
Boiling Point 245°C
Quantity 5 g
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Indanone