accessibility menu, dialog, popup

UserName

O

CAS RN 83-33-0

IUPAC Name: 2,3-dihydro-1H-inden-1-one
Synonyms: indanone hydrindone 1-indone α-indanone α-hydrindone
Molecular Weight (g/mol): 132.16
Molecular Formula: C9H8O
InChi Key: QNXSIUBBGPHDDE-UHFFFAOYSA-N
SMILES: O=C1CCC2=CC=CC=C12
Boiling Point 
  • (4)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (2)
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (4)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)