missing translation for 'onlineSavingsMsg'
Learn More

(1-Imidazolyl)acetonitrile 98.0+%, TCI America™

Catalog No. I09905G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09905G 5 g
I09901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09905G Supplier TCI America Supplier No. I09905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98873-55-3
Molecular Formula C5H5N3
Molecular Weight (g/mol) 107.116
MDL Number MFCD06421433
InChI Key ZPGCVVBPGQJSPX-UHFFFAOYSA-N
Synonym 2-1h-imidazol-1-yl acetonitrile, imidazol-1-yl-acetonitrile, 1-cyanomethyl imidazole, 1h-imidazole-1-acetonitrile, 1h-imidazol-1-ylacetonitrile, 2-1-imidazolyl acetonitrile, 1-imidazolyl acetonitrile, 2-imidazol-1-yl acetonitrile, 1h-imidazol-1-yl acetonitrile, 1-cyanomethylimidazole
PubChem CID 11804695
IUPAC Name 2-imidazol-1-ylacetonitrile
SMILES C1=CN(C=N1)CC#N

Specifications

Melting Point 56°C
Quantity 5 g
UN Number 3439
Formula Weight 107.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1-Imidazolyl)acetonitrile