missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxypyrene 98.0+%, TCI America™

Catalog No. p-7131043
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14351G 1 g
H14355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14351G Supplier TCI America Supplier No. H14351G
Only null left

Chemical Identifiers

CAS 5315-79-7
Molecular Formula C16H10O
Molecular Weight (g/mol) 218.255
MDL Number MFCD00044543
InChI Key BIJNHUAPTJVVNQ-UHFFFAOYSA-N
Synonym 1-hydroxypyrene, 1-pyrenol, 3-hydroxypyrene, pyrenol, unii-n2h6o5v707, hydroxypyrene, 1-hydroxy-pyrene, 3-pyrenol, 1-pyrenol 8ci
PubChem CID 21387
ChEBI CHEBI:34093
IUPAC Name pyren-1-ol
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)O

Specifications

Melting Point 180°C
Quantity 1 g
Formula Weight 218.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hydroxypyrene
RTECSNumber UR2845000
TSCA No