missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxypyrene, 99+%

Catalog No. AC232272500
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC232272500 250 mg Glass bottle
AC232270500 50 mg Glass bottle
AC232270010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC232272500 Supplier Thermo Scientific Chemicals Supplier No. 232272500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5315-79-7 | C16H10O | 218.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5315-79-7
Molecular Formula C16H10O
Molecular Weight (g/mol) 218.25
MDL Number MFCD00044543
InChI Key BIJNHUAPTJVVNQ-UHFFFAOYSA-N
Synonym 1-hydroxypyrene, 1-pyrenol, 3-hydroxypyrene, pyrenol, unii-n2h6o5v707, hydroxypyrene, 1-hydroxy-pyrene, 3-pyrenol, 1-pyrenol 8ci
PubChem CID 21387
ChEBI CHEBI:34093
IUPAC Name pyren-1-ol
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 176°C to 178°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in most common organic solvents
Formula Weight 218.25
Percent Purity 99+%
Physical Form Powder or Fine Needles
Chemical Name or Material 1-Hydroxypyrene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only