missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxyindan 99.0+%, TCI America™

Catalog No. p-7130577
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05165G 5 g
H051625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05165G Supplier TCI America Supplier No. H05165G
Only null left

Chemical Identifiers

CAS 10-6-6351
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00003797
InChI Key YIAPLDFPUUJILH-UHFFFAOYNA-N
Synonym 1-indanol, indanol, indan-1-ol, 1-hydroxyhydrindene, 1h-inden-1-ol, 2,3-dihydro, 1h-indenol, 2,3-dihydro, 1-hydroxyindan, 1-hydroxyindane, 1-indanole, --indanol
PubChem CID 22819
ChEBI CHEBI:16697
IUPAC Name 2,3-dihydro-1H-inden-1-ol
SMILES C1CC2=CC=CC=C2C1O

Specifications

Melting Point 52°C
Color White-Yellow
Quantity 5 g
Formula Weight 134.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hydroxyindan
RTECSNumber NK7300000
TSCA No