missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxyethylethoxypiperazine, 95%

CAS: 13349-82-1 | C8H18N2O2 | 174.24 g/mol

$104.20 - $344.85

Chemical Identifiers

CAS 13349-82-1
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.24
MDL Number MFCD00074931
InChI Key FLNQAPQQAZVRDA-UHFFFAOYSA-N
Synonym 1-2-2-hydroxyethoxy ethyl piperazine, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, 2-2-1-piperazinyl ethoxy ethanol, 1-2-2-hydroxyethoxyl-ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, ethanol, 2-2-1-piperazinyl ethoxy
PubChem CID 139436
IUPAC Name 2-(2-piperazin-1-ylethoxy)ethanol
SMILES C1CN(CCN1)CCOCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC259110050
SDS View Documents
Thermo Scientific Chemicals
259110050
5 mL Glass bottle
Each for $104.20
Only null left
 
AC259110250
SDS View Documents
Thermo Scientific Chemicals
259110250
25 mL Glass bottle
Each for $344.85
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13349-82-1
174.24
FLNQAPQQAZVRDA-UHFFFAOYSA-N
139436
C1CN(CCN1)CCOCCO
C8H18N2O2
MFCD00074931
1-2-2-hydroxyethoxy ethyl piperazine, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, 2-2-1-piperazinyl ethoxy ethanol, 1-2-2-hydroxyethoxyl-ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, ethanol, 2-2-1-piperazinyl ethoxy
2-(2-piperazin-1-ylethoxy)ethanol

Specifications

13349-82-1
100.0
1.0100g/mL
Authentic
Glass bottle
1.4960 to 1.4980
5 mL
1-2-2-hydroxyethoxy ethyl piperazine, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, 2-2-1-piperazinyl ethoxy ethanol, 1-2-2-hydroxyethoxyl-ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, ethanol, 2-2-1-piperazinyl ethoxy
C1CN(CCN1)CCOCCO
174.24
174.24
Liquid
94.0
Brown-Yellow to Colorless
150.0°C (13.0 mmHg)
94% min. (GC)
C8H18N2O2
MFCD00074931
1.01
FLNQAPQQAZVRDA-UHFFFAOYSA-N
2-(2-piperazin-1-ylethoxy)ethanol
139436
95%
1-Hydroxyethylethoxypiperazine
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only