missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hydroxyethylethoxypiperazine, 95%
CAS: 13349-82-1 | C8H18N2O2 | 174.24 g/mol
Supplier: Thermo Scientific Chemicals 259110250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Hydroxyethylethoxypiperazine | |
| 94.0 | |
| Brown-Yellow to Colorless | |
| 150.0°C (13.0 mmHg) | |
| 94% min. (GC) | |
| C8H18N2O2 | |
| MFCD00074931 | |
| 1.01 | |
| FLNQAPQQAZVRDA-UHFFFAOYSA-N | |
| 2-(2-piperazin-1-ylethoxy)ethanol | |
| 139436 | |
| 95% |
| 13349-82-1 | |
| 100.0 | |
| 1.0100g/mL | |
| Authentic | |
| Glass bottle | |
| 1.4960 to 1.4980 | |
| 25 mL | |
| 1-2-2-hydroxyethoxy ethyl piperazine, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, 2-2-1-piperazinyl ethoxy ethanol, 1-2-2-hydroxyethoxyl-ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, ethanol, 2-2-1-piperazinyl ethoxy | |
| C1CN(CCN1)CCOCCO | |
| 174.24 | |
| 174.24 | |
| Liquid |
Chemical Identifiers
| 13349-82-1 | |
| 174.24 | |
| FLNQAPQQAZVRDA-UHFFFAOYSA-N | |
| 139436 | |
| C1CN(CCN1)CCOCCO |
| C8H18N2O2 | |
| MFCD00074931 | |
| 1-2-2-hydroxyethoxy ethyl piperazine, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, 2-2-1-piperazinyl ethoxy ethanol, 1-2-2-hydroxyethoxyl-ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, ethanol, 2-2-1-piperazinyl ethoxy | |
| 2-(2-piperazin-1-ylethoxy)ethanol |
RUO – Research Use Only