missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxybenzotriazole Monohydrate 97.0+%, TCI America™

Catalog No. H046825G
Change view
Click to view available options
Quantity:
25 g
200 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H046825G 25 g
H0468200G 200 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H046825G Supplier TCI America Supplier No. H046825G

Chemical Identifiers

CAS 80029-43-2
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00065690
InChI Key PJUPKRYGDFTMTM-UHFFFAOYSA-N
Synonym 1,2,3-Benzotriazol-1-ol, HOBt
PubChem CID 2796029
IUPAC Name 1H-1,2,3-benzotriazol-1-ol hydrate
SMILES O.ON1N=NC2=CC=CC=C12

Specifications

Melting Point 160°C
Color White
Quantity 25 g
UN Number 3474
Formula Weight 135.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hydroxybenzotriazole Monohydrate
EINECSNumber (5)-3579&(5)-3603
RTECSNumber DM1288000
TSCA No