accessibility menu, dialog, popup

UserName

NNNHOH2O

CAS RN 80029-43-2

IUPAC Name: 1H-1,2,3-benzotriazol-1-ol hydrate
Synonyms: hydroxybenzotriazole hydrate
Molecular Weight (g/mol): 153.14
Molecular Formula: C6H7N3O2
InChi Key: PJUPKRYGDFTMTM-UHFFFAOYSA-N
SMILES: O.ON1N=NC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results