missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hydroxyanthraquinone 98.0+%, TCI America™
$106.25 - $318.75
Chemical Identifiers
| CAS | 129-43-1 |
|---|---|
| Molecular Formula | C14H8O3 |
| Molecular Weight (g/mol) | 224.215 |
| MDL Number | MFCD00058946 |
| InChI Key | BTLXPCBPYBNQNR-UHFFFAOYSA-N |
| PubChem CID | 8512 |
| ChEBI | CHEBI:28877 |
| IUPAC Name | 1-hydroxyanthracene-9,10-dione |
| SMILES | C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)O |
Chemical Identifiers
| 129-43-1 | |
| 224.215 | |
| BTLXPCBPYBNQNR-UHFFFAOYSA-N | |
| CHEBI:28877 | |
| C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)O |
| C14H8O3 | |
| MFCD00058946 | |
| 8512 | |
| 1-hydroxyanthracene-9,10-dione |
Specifications
| 129-43-1 | |
| C14H8O3 | |
| 5 g | |
| C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)O | |
| 224.215 | |
| CHEBI:28877 | |
| ≥98.0% (HPLC) | |
| 1-Hydroxyanthraquinone |
| 198°C | |
| MFCD00058946 | |
| BTLXPCBPYBNQNR-UHFFFAOYSA-N | |
| 1-hydroxyanthracene-9,10-dione | |
| 8512 | |
| 224.22 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0703
RTECSNumber : CB7178000
TSCA : Yes