accessibility menu, dialog, popup

UserName

CAS RN 129-43-1

IUPAC Name: 1-hydroxy-9,10-dihydroanthracene-9,10-dione
Synonyms: hydroxyanthraquinone α-hydroxyanthraquinone 1-hydroxyanthra-9,10-quinone
Molecular Weight (g/mol): 224.22
Molecular Formula: C14H8O3
InChi Key: BTLXPCBPYBNQNR-UHFFFAOYSA-N
SMILES: OC1=CC=CC2=C1C(=O)C1=CC=CC=C1C2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results