missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxy-6-(trifluoromethyl)benzotriazole 98.0+%, TCI America™

Catalog No. p-7131053
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14541G 1 g
H14545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14541G Supplier TCI America Supplier No. H14541G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26198-21-0
Molecular Formula C7H4F3N3O
Molecular Weight (g/mol) 203.124
MDL Number MFCD00038806
InChI Key DGIBHCWBCOAPDN-UHFFFAOYSA-N
PubChem CID 688194
IUPAC Name 1-hydroxy-6-(trifluoromethyl)benzotriazole
SMILES C1=CC2=C(C=C1C(F)(F)F)N(N=N2)O

Specifications

Melting Point 148°C
Color White
Quantity 1 g
UN Number 1325
Formula Weight 203.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hydroxy-6-(trifluoromethyl)benzotriazole
EINECSNumber (5)-3578
TSCA No