accessibility menu, dialog, popup

UserName

CAS RN 26198-21-0

IUPAC Name: 6-(trifluoromethyl)-1H-1,2,3-benzotriazol-1-ol
Synonyms: 6-(trifluoromethyl)-1,2,3-benzotriazol-1-ol
Molecular Weight (g/mol): 203.12
Molecular Formula: C7H4F3N3O
InChi Key: DGIBHCWBCOAPDN-UHFFFAOYSA-N
SMILES: ON1N=NC2=CC=C(C=C12)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results