missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxy-2-naphthaldehyde 98.0+%, TCI America™

Catalog No. H08391G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08391G 1 g
H08395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08391G Supplier TCI America Supplier No. H08391G
Only null left

Chemical Identifiers

CAS 574-96-9
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
MDL Number MFCD00092325
InChI Key OITQDWKMIPXGFL-UHFFFAOYSA-N
Synonym 1-Naphthol-2-carboxaldehyde
PubChem CID 443195
ChEBI CHEBI:28091
IUPAC Name 1-hydroxynaphthalene-2-carbaldehyde
SMILES C1=CC=C2C(=C1)C=CC(=C2O)C=O

Specifications

Melting Point 56°C
Color Green-Yellow
Quantity 1 g
Formula Weight 172.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hydroxy-2-naphthaldehyde