missing translation for 'onlineSavingsMsg'
Learn More

1'-Hydroxy-2'-acetonaphthone 98.0+%, TCI America™

Catalog No. p-7130815
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
H093525G 25 g
H0935100G 100 g
2 options

Catalog No. H093525G

Supplier: TCI America H093525G

Only null left

Chemical Identifiers

CAS 711-79-5
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00003963
InChI Key JBGJVMVWYWUVOW-UHFFFAOYSA-N
Synonym 1'-hydroxy-2'-acetonaphthone, 2-acetyl-1-naphthol, 1-hydroxy-2-acetylnaphthalene, 2-acetyl-1-hydroxynaphthalene, 1-hydroxy-2-acetonaphthone, ethanone, 1-1-hydroxy-2-naphthalenyl, 1-1-hydroxynaphthalen-2-yl ethanone, 1-1-hydroxy-2-naphthalenyl ethanone, 2'-acetonaphthone, 1'-hydroxy, 1-hydroxy-2-naphthyl methyl ketone
PubChem CID 69733
IUPAC Name 1-(1-hydroxynaphthalen-2-yl)ethan-1-one
SMILES CC(=O)C1=CC=C2C=CC=CC2=C1O

Specifications

Melting Point 99°C
Quantity 25 g
Formula Weight 186.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1′-Hydroxy-2′-acetonaphthone