missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyne 97.0+%, TCI America™

Catalog No. p-7130366
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H014025ML 25 mL
H0140250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H014025ML Supplier TCI America Supplier No. H014025ML
Only null left

Chemical Identifiers

CAS 693-02-7
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00009504
InChI Key CGHIBGNXEGJPQZ-UHFFFAOYSA-N
Synonym 1-hexyne, n-butylacetylene, butylacetylene, hexyne, unii-5fzf2f38f5, n-hexyne, 1-hexyn, hex-5-yn-1-yl, pubchem13057
PubChem CID 12732
IUPAC Name hex-1-yne
SMILES CCCCC#C

Specifications

Color Yellow
Boiling Point 71°C
Quantity 25 mL
UN Number 3295
Formula Weight 82.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Hexyne
RTECSNumber MQ9691000
TSCA Yes